Learn More
Medchemexpress LLC LX-1031 | 945976-76-1 | 99.14% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1304110MM/1ML
Encompass_Preferred
LX-1031 is a potent, orally available tryptophan 5-hydroxylase (TPH) inhibitor that reduces serotonin (5-HT) synthesis peripherally.
  • Potent and orally available: effective when administered orally.
  • Peripheral serotonin reduction: specifically reduces serotonin (5-HT) synthesis in the periphery, without affecting brain 5-HT levels.
  • Reversible effects: in mice, the effects on jejunal mucosa 5-HT levels were reversible within 2 days of discontinuation.
  • Research use only: this product is intended for research use only and is not for sale to patients.
  • Clinical trial information: has been investigated in a Phase 2 clinical trial (NCT00813098) for Irritable Bowel Syndrome.
Catalog No. 50-003-38304