Learn More
TARGETMOL CHEMICALS INC LX-6171 5MG
SDP
Catalog No. 502580560
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LX-6171 a proline transporter (SLC6A7) inhibitor is used potentially for the treatment of schizophrenia and Alzheimer's disease. purity: 97%
Catalog No. 50-258-0560 Supplier TARGETMOL CHEMICALS INC Supplier No. T278655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LX-6171 a proline transporter (SLC6A7) inhibitor is used potentially for the treatment of schizophrenia and Alzheimer's disease. purity: 97%