Learn More
Electron Microscopy Sciences ProScope LX-Proscope HR
SDP
Catalog No. 5033677
Encompass
Capture stills, video, time lapse, perform detailed measurement, and annotate your still images. Works with original ProScope or ProScope HR under Windows XP SP2/Vista and up and Mac OS X 10.4.8 and up.
Catalog No. 50-336-77 Supplier Electron Microscopy Sciences Supplier No. 6835322
Capture stills, video, time lapse, perform detailed measurement, and annotate your still images. Works with original ProScope or ProScope HR under Windows XP SP2/Vista and up and Mac OS X 10.4.8 and up.