Learn More
Ladd Research Industries Inc LX112 RESIN 450ML
SDP
Catalog No. NC9925769
Encompass
Direct replacement for Epon 812 in all formulations. Infiltration, embedding, sectioning and preservation of tissue is identical to Epon 812.
Catalog No. NC9925769 Supplier Ladd Research Industries Inc Supplier No. 21310
Direct replacement for Epon 812 in all formulations. Infiltration, embedding, sectioning and preservation of tissue is identical to Epon 812.