Learn More
Medchemexpress LLC HY-111383 5mg Medchemexpress, LX2343 CAS:333745-53-2 Purity:>98%
SDP
Catalog No. 501968848
Encompass_Preferred
Medchemexpress, HY-111383 5mg LX2343 CAS:333745-53-2 LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8848 Supplier Medchemexpress LLC Supplier No. HY1113835MG
Medchemexpress, HY-111383 5mg LX2343 CAS:333745-53-2 LX2343 is a BACE1 enzyme inhibitor with an IC50 value of 11.43±0.36 μM. LX2343 acts as a non-ATP competitive PI3K inhibitor with an IC50 of 15.99±3.23 μM. LX2343 stimulates autophagy in its promotion of Aβ clearance. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.