Learn More
TA Instruments LX500 365nm UV Head V3 Kit, Designed for LX500 Controller, High Output UV LEDs, Higher Power Delivery, Improved Efficiency, Enhanced Beam Profile, Industrial Manufacturing Applications
Catalog No. 5000580756 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments LX500 365nm UV Head V3 Kit is designed for use with the OmniCure LX500 Controller. The V3 UV LED Heads feature next generation high output UV LEDs that deliver higher power, improved efficiency, and an enhanced beam profile compared to earlier versions. The heads are optimized for industrial manufacturing applications with cross compatible accessories available. They deliver a high dosage distributed uniformly across the spot area, enabling faster, more consistent cures with greater control and efficiency.
Catalog No. 50-005-80756 Supplier TA Instruments Supplier No. 974022.901
Waters TA Instruments LX500 365nm UV Head V3 Kit is designed for use with the OmniCure LX500 Controller. The V3 UV LED Heads feature next generation high output UV LEDs that deliver higher power, improved efficiency, and an enhanced beam profile compared to earlier versions. The heads are optimized for industrial manufacturing applications with cross compatible accessories available. They deliver a high dosage distributed uniformly across the spot area, enabling faster, more consistent cures with greater control and efficiency.