Learn More
TA Instruments LX500 385nm UV Head V3 Kit, Designed for LX500 Controller, High Output UV LEDs, Higher Power Delivery, Improved Efficiency, Enhanced Beam Profile, Industrial Manufacturing Applications
Catalog No. 5000580676 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments LX500 385nm UV Head V3 Kit is designed for use with the OmniCure LX500 Controller. The V3 UV LED Heads feature high output UV LEDs for best-in-class optical performance. They deliver higher power, improved efficiency, and an enhanced beam profile over earlier models. Optimized for use in industrial manufacturing, the heads also support a wide range of compatible accessories. The V3 Heads deliver uniform dosage across the spot area for faster and more consistent cures with improved efficiency and control.
Catalog No. 50-005-80676 Supplier TA Instruments Supplier No. 974023.901
Waters TA Instruments LX500 385nm UV Head V3 Kit is designed for use with the OmniCure LX500 Controller. The V3 UV LED Heads feature high output UV LEDs for best-in-class optical performance. They deliver higher power, improved efficiency, and an enhanced beam profile over earlier models. Optimized for use in industrial manufacturing, the heads also support a wide range of compatible accessories. The V3 Heads deliver uniform dosage across the spot area for faster and more consistent cures with improved efficiency and control.