Learn More
TA Instruments LX500 405nm UV Head V3 Kit, Designed for LX500 Controller, High Output UV LEDs, Higher Power Delivery, Improved Efficiency, Enhanced Beam Profile, Industrial Manufacturing Applications
Catalog No. 5000580692 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments LX500 405nm UV Head V3 Kit is designed for use with the OmniCure LX500 Controller. The V3 UV LED Heads incorporate advanced high output LEDs to deliver improved power, efficiency, and beam profile. Optimized for industrial manufacturing applications, the heads are compatible with multiple accessories. They provide a high, uniformly distributed dosage for faster cures, consistent performance, and better process control.
Catalog No. 50-005-80692 Supplier TA Instruments Supplier No. 974025.901
Waters TA Instruments LX500 405nm UV Head V3 Kit is designed for use with the OmniCure LX500 Controller. The V3 UV LED Heads incorporate advanced high output LEDs to deliver improved power, efficiency, and beam profile. Optimized for industrial manufacturing applications, the heads are compatible with multiple accessories. They provide a high, uniformly distributed dosage for faster cures, consistent performance, and better process control.