Learn More
Invitrogen™ LXA4 Competitive ELISA Kit
Description
The multispecies Lipoxin A4 (LXA4) ELISA quantitates LXA4 in serum, plasma, and other biological fluids Principle of the method: The Competitive multispecies ELISA research-use-only kit is designed to quantitatively measure Lipoxin A4 (LXA4) independent of species. A LXA4 standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to LXA4is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of LXA4 in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Assay Range | 0.78-50 ng/mL |
| Assay Sensitivity | 0.47 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Protein Family | Specialized pro-resolving mediators |
| Sample Type | Plasma, Serum |
| Target Species | All Species |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | Lipoxin A4Lipoxin A4; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.