Learn More
Medchemexpress LLC HY-112089 10mg , LXH254 CAS:1800398-38-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11208910MG
Encompass_Preferred
HY-112089 10mg LXH254 CAS: 1800398-38-2 LXH254 is a potent, selective, orally active, type II BRAF and CRAF inhibitor, with IC50 values of 0.072 and 0.21 nM against CRAF and BRAF, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9939