Learn More
Selleck Chemical LLC LXH254 2mg 1800398-38-2
SDP
Catalog No. 501948174
Encompass_Preferred
LXH254 is a type II ATP-competitive inhibitor that inhibits both B- and CRAF kinase activities at picomolar concentrations with a high degree of selectivity against a panel of 456 human kinases and in cell-based assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8174 Supplier Selleck Chemical LLC Supplier No. S87452MG
LXH254 is a type II ATP-competitive inhibitor that inhibits both B- and CRAF kinase activities at picomolar concentrations with a high degree of selectivity against a panel of 456 human kinases and in cell-based assays. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.