Learn More
TARGETMOL CHEMICALS INC LXH254 5MG
SDP
Catalog No. 502577437
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LXH254 is a potent C-Raf and B-Raf inhibitor. purity: 99%
Catalog No. 50-257-7437 Supplier TARGETMOL CHEMICALS INC Supplier No. T118985MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LXH254 is a potent C-Raf and B-Raf inhibitor. purity: 99%