Learn More
Aviva Systems Biology Corp LXN Antibody - N-terminal region (ARP65175_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65175_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LXN. It was validated on Western Blot. . Alternate names: ECI, TCIApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-449