Learn More
Selleck Chemical LLC LXR-623
SDP
Catalog No. 5000000672
Encompass_Preferred
LXR-623 is a novel liver X-receptor(LXR) agonist with IC50 values of 179 nM and 24 nM for LXR- and LXR- respectively It is orally bioavailable and readily passes the blood-brain barrier
Catalog No. 50-000-00672 Supplier Selleck Chemical LLC Supplier No. S8390200MG
LXR-623 is a novel liver X-receptor(LXR) agonist with IC50 values of 179 nM and 24 nM for LXR- and LXR- respectively It is orally bioavailable and readily passes the blood-brain barrier