Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LXR-α protein, human | 95.0% | 10UG
SDP

Catalog No. 5000176181
Encompass_Preferred

Recombinant human LXR-α protein (NR1H3), corresponding to UniProt Q13133 residues Q182-E447, expressed in Escherichia coli and supplied as a purified, tag-free solution in 25 mM Tris-HCl, 50 mM NaCl, pH 8.5. Reported concentration is 8.58 mg/mL, molecular weight ≈30.7 kDa, and purity ≥95% by reducing SDS-PAGE. Store at -80°C and avoid repeated freeze-thaw cycles. For research use only.

  • High purity validated by reducing SDS-PAGE.
  • Tag-free construct for unaltered receptor properties.
  • Defined sequence range (Q182-E447) for domain-focused studies.
  • Supplied as a 0.22 μm filtered solution for immediate use.
  • Low endotoxin suitable for biochemical assays.
  • Stable when stored at -80°C and shipped on dry ice.

Catalog No. 50-001-76181 Supplier Medchemexpress LLC Supplier No. HYP70140410UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human LXR-α protein (NR1H3), corresponding to UniProt Q13133 residues Q182-E447, expressed in Escherichia coli and supplied as a purified, tag-free solution in 25 mM Tris-HCl, 50 mM NaCl, pH 8.5. Reported concentration is 8.58 mg/mL, molecular weight ≈30.7 kDa, and purity ≥95% by reducing SDS-PAGE. Store at -80°C and avoid repeated freeze-thaw cycles. For research use only.

  • High purity validated by reducing SDS-PAGE.
  • Tag-free construct for unaltered receptor properties.
  • Defined sequence range (Q182-E447) for domain-focused studies.
  • Supplied as a 0.22 μm filtered solution for immediate use.
  • Low endotoxin suitable for biochemical assays.
  • Stable when stored at -80°C and shipped on dry ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.