Learn More
MSC Makita Chain Saw, 36V, Brushless, 16 in Bar, 1 Kit
Catalog No. 502816393
Encompass
This 36V (18V X2) LXT Brushless 16 in Chain Saw kit is a cordless alternative to gas-powered outdoor power equipment, offering zero emissions, lower noise, and reduced maintenance. Powered by two 18V LXT 4.0Ah batteries, it delivers maximum power, speed, and run-time for cutting and trimming tasks.
  • Brushless motor direct-drive system delivers power comparable to a 32cc gas chain saw
  • Tool-less chain adjustment
  • Variable speed trigger with 0-3,940 FPM chain speed
  • Adjustable automatic chain lubrication
  • Front hand guard with chain brake and electric brake
  • Extreme Protection Technology (XPT) for increased dust and water resistance
  • Star Protection Computer Controls protects against overloading and overheating
Catalog No. 50-281-6393 Supplier MSC Supplier No. 06302194
This 36V (18V X2) LXT Brushless 16 in Chain Saw kit is a cordless alternative to gas-powered outdoor power equipment, offering zero emissions, lower noise, and reduced maintenance. Powered by two 18V LXT 4.0Ah batteries, it delivers maximum power, speed, and run-time for cutting and trimming tasks.
  • Brushless motor direct-drive system delivers power comparable to a 32cc gas chain saw
  • Tool-less chain adjustment
  • Variable speed trigger with 0-3,940 FPM chain speed
  • Adjustable automatic chain lubrication
  • Front hand guard with chain brake and electric brake
  • Extreme Protection Technology (XPT) for increased dust and water resistance
  • Star Protection Computer Controls protects against overloading and overheating