Learn More
MSC Makita Power Head, 36V LXT, Brushless, String Trimmer, 1 ea
Catalog No. 502819097
Encompass
This 36V (18V X2) LXT® Brushless Couple Shaft Power Head with String Trimmer Attachment offers a cordless alternative to gas-powered outdoor power equipment, providing zero emissions, lower noise, and maximum versatility. It is powered by two 18V LXT® batteries (sold separately) and is compatible with various optional couple shaft attachments.
  • Brushless motor delivers up to 7,100 RPM
  • Offers 3-speed options (4,200, 6,000, 7,100 RPM)
  • Variable speed trigger for controlled cutting speed
  • Compatible with multiple couple shaft attachments
  • Tool-less attachment installation and replacement
  • Reverse rotation system clears entangled grass and weeds
  • Extreme Protection Technology (XPT™) for dust and water resistance
  • Star Protection Computer Controls™ protects against overloading
  • Includes 3-year limited warranty
Catalog No. 50-281-9097 Supplier MSC Supplier No. 15596356
This 36V (18V X2) LXT® Brushless Couple Shaft Power Head with String Trimmer Attachment offers a cordless alternative to gas-powered outdoor power equipment, providing zero emissions, lower noise, and maximum versatility. It is powered by two 18V LXT® batteries (sold separately) and is compatible with various optional couple shaft attachments.
  • Brushless motor delivers up to 7,100 RPM
  • Offers 3-speed options (4,200, 6,000, 7,100 RPM)
  • Variable speed trigger for controlled cutting speed
  • Compatible with multiple couple shaft attachments
  • Tool-less attachment installation and replacement
  • Reverse rotation system clears entangled grass and weeds
  • Extreme Protection Technology (XPT™) for dust and water resistance
  • Star Protection Computer Controls™ protects against overloading
  • Includes 3-year limited warranty