Learn More
MSC Makita Cordless Stick Vacuum, 18V LXT Brushless, Trigger, 1 kit
Catalog No. 502816123
Encompass
The Makita 18V LXT Brushless Cordless Vacuum Kit offers compact, powerful cleaning with strong suction. It features both trigger and slide switch modes for convenience and a bagless cloth filtration system for easy debris disposal.
  • BL Brushless Motor delivers up to 49 CFM.
  • Provides up to 13 minutes of continuous use from a 2.0Ah battery.
  • Bagless cloth filtration for easy cleaning and debris disposal.
  • Strong suction power for faster, efficient cleaning.
  • Operates with trigger or slide switch mode.
  • Compact, ergonomic design, weighing 2.7 lbs. with battery, reduces fatigue.
  • Increased storage capacity for longer intervals between disposal.
  • Ergonomic soft grip handle for comfort and control.
  • Flat nozzle with ball-and-socket feature for maneuverability.
Catalog No. 50-281-6123 Supplier MSC Supplier No. 06299978
The Makita 18V LXT Brushless Cordless Vacuum Kit offers compact, powerful cleaning with strong suction. It features both trigger and slide switch modes for convenience and a bagless cloth filtration system for easy debris disposal.
  • BL Brushless Motor delivers up to 49 CFM.
  • Provides up to 13 minutes of continuous use from a 2.0Ah battery.
  • Bagless cloth filtration for easy cleaning and debris disposal.
  • Strong suction power for faster, efficient cleaning.
  • Operates with trigger or slide switch mode.
  • Compact, ergonomic design, weighing 2.7 lbs. with battery, reduces fatigue.
  • Increased storage capacity for longer intervals between disposal.
  • Ergonomic soft grip handle for comfort and control.
  • Flat nozzle with ball-and-socket feature for maneuverability.