Learn More
MSC Makita Material Handler, 36V LXT Brushless, Flat Bucket, 1 ea
Catalog No. 502817624
Encompass
The Makita XUC06Z is a 36V (18V X2) LXT Brushless Material Handler equipped with a flat bucket, designed for efficient material transport on job sites and other environments.
  • Operates on two 18V LXT lithium-ion batteries, with capacity for four for extended runtime up to 100 minutes.
  • A 295 L heaped capacity and a maximum load of 661 lbs.
  • Three forward speeds and one reverse speed, with variable control.
  • Flat bucket features a capped nozzle for liquid draining.
  • Mechanical axle disc brake and electromagnetic brake for enhanced safety.
  • Integrated headlights, taillight, beeper, and level gauge.
  • Adjustable rubberized soft grip handles and brushless motor for optimal performance.
  • Extreme Protection Technology (XPT™) provides dust and water resistance.
Catalog No. 50-281-7624 Supplier MSC Supplier No. 06302145
Add to Cart
Add to Cart
Request a Quote
The Makita XUC06Z is a 36V (18V X2) LXT Brushless Material Handler equipped with a flat bucket, designed for efficient material transport on job sites and other environments.
  • Operates on two 18V LXT lithium-ion batteries, with capacity for four for extended runtime up to 100 minutes.
  • A 295 L heaped capacity and a maximum load of 661 lbs.
  • Three forward speeds and one reverse speed, with variable control.
  • Flat bucket features a capped nozzle for liquid draining.
  • Mechanical axle disc brake and electromagnetic brake for enhanced safety.
  • Integrated headlights, taillight, beeper, and level gauge.
  • Adjustable rubberized soft grip handles and brushless motor for optimal performance.
  • Extreme Protection Technology (XPT™) provides dust and water resistance.