Learn More
MSC Makita String Trimmer, 40V max, Brushless, 17 in, 1 ea
Catalog No. 502819116
Encompass
The Makita CRU03Z is a 40V max ConnectX™ brushless string trimmer designed for professional landscaping. It delivers power equivalent to a 30cc gas string trimmer with zero emissions and reduced maintenance. Featuring an external battery connection, it offers improved balance and extended run time for all-day use. This model is 'Tool Only' and does not include a battery.
  • Power equivalent to a 30cc gas string trimmer
  • Provides a 17 in cutting swath for increased productivity
  • External battery connection shifts weight for improved balance
  • Steel drive shaft reduces vibration
  • Automatic Torque Drive Technology adjusts speed from 4,000 to 6,000 RPM
  • Three speed options for power management
  • Reverse rotation function clears entangled grass and weeds
  • Extreme Protection Technology (XPT™) provides dust and water resistance
Catalog No. 50-281-9116 Supplier MSC Supplier No. 15596554
The Makita CRU03Z is a 40V max ConnectX™ brushless string trimmer designed for professional landscaping. It delivers power equivalent to a 30cc gas string trimmer with zero emissions and reduced maintenance. Featuring an external battery connection, it offers improved balance and extended run time for all-day use. This model is 'Tool Only' and does not include a battery.
  • Power equivalent to a 30cc gas string trimmer
  • Provides a 17 in cutting swath for increased productivity
  • External battery connection shifts weight for improved balance
  • Steel drive shaft reduces vibration
  • Automatic Torque Drive Technology adjusts speed from 4,000 to 6,000 RPM
  • Three speed options for power management
  • Reverse rotation function clears entangled grass and weeds
  • Extreme Protection Technology (XPT™) provides dust and water resistance