Learn More
Enzo Life Sciences LY 171883 (10 mg). CAS: 88107-10-2
SDP
Supplier:  Enzo Life Sciences BMLRA1010010
Encompass_Preferred
Selective, orally active leukotriene D4 antagonist. Has been shown to bind to the γ-isoform of the peroxisome proliferator-activated receptor (PPARγ). Alternative name: Tomelukast. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (25mg/ml) or dimethyl formamide. Also soluble in 0.5M aqueous sodium bicarbonate. Long Term Storage: Ambient.
Catalog No. 50-201-1703