Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ly 2389575 hydrochloride | 885104-09-6 | MFCD28133385 | 99.9% | 438.58 g/mol | C15H16BrCl3N4 | 100 MG
SDP

Catalog No. 5000241448
Encompass_Preferred

LY2389575 hydrochloride is a selective, noncompetitive negative allosteric modulator (NAM) of the metabotropic glutamate receptor 3 (mGlu3) supplied as the hydrochloride salt for research use. It is provided as a solid for in vitro and preclinical studies, including investigations of amyloid-β toxicity.

  • Selective noncompetitive mGlu3 negative allosteric modulator.
  • Supplied as hydrochloride salt in solid form, white to light yellow.
  • High purity: 99.9%.
  • Molecular formula C15H16BrCl3N4; molecular weight 438.58 g/mol.
  • Available in multiple pack sizes and as a DMSO solution for convenience.
  • Recommended storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C.

Catalog No. 50-002-41448 Supplier Medchemexpress LLC Supplier No. HY107509100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LY2389575 hydrochloride is a selective, noncompetitive negative allosteric modulator (NAM) of the metabotropic glutamate receptor 3 (mGlu3) supplied as the hydrochloride salt for research use. It is provided as a solid for in vitro and preclinical studies, including investigations of amyloid-β toxicity.

  • Selective noncompetitive mGlu3 negative allosteric modulator.
  • Supplied as hydrochloride salt in solid form, white to light yellow.
  • High purity: 99.9%.
  • Molecular formula C15H16BrCl3N4; molecular weight 438.58 g/mol.
  • Available in multiple pack sizes and as a DMSO solution for convenience.
  • Recommended storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.