Learn More
Medchemexpress LLC Ly-2584702 tosylate salt | 1082949-68-5 | 99.5% | 617.62 | C28H27F4N7O3S | 100MG
SDP
Supplier:  Medchemexpress LLC HY12493A100MG
Encompass_Preferred
LY-2584702 tosylate salt is the tosylate salt form of LY-2584702, a selective ATP-competitive inhibitor of p70S6K (S6K1) used in biochemical and cellular research to inhibit S6K signaling and downstream phosphorylation of the S6 ribosomal protein. It exhibits potent enzyme inhibition in the low nanomolar range and cellular activity in the submicromolar range, and is supplied in solid and solution formats for assay use.
  • Selective ATP-competitive p70S6K inhibitor.
  • Potent enzyme inhibition with low-nanomolar IC50 values.
  • Demonstrated cellular inhibition of pS6 at submicromolar concentrations.
  • Available in multiple solid sizes and a DMSO solution for assay preparation.
  • High reported purity suitable for biochemical assays.
  • Provided as tosylate salt to aid handling and formulation.
Catalog No. 50-002-36035