Learn More
Medchemexpress LLC HY-12493A 5mg Medchemexpress, LY-2584702 (tosylate salt) CAS:1082949-68-5 Purity:>98%
SDP
Catalog No. 502030014
Encompass_Preferred
Medchemexpress, HY-12493A 5mg LY-2584702 (tosylate salt) CAS:1082949-68-5 LY-2584702 tosylate salt is a selective ATP competitive inhibitor of p70S6K with an IC50 of 4 nM. In S6K1 enzyme assay, the IC50 of LY-2584702 is 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0014 Supplier Medchemexpress LLC Supplier No. HY12493A5MG
Medchemexpress, HY-12493A 5mg LY-2584702 (tosylate salt) CAS:1082949-68-5 LY-2584702 tosylate salt is a selective ATP competitive inhibitor of p70S6K with an IC50 of 4 nM. In S6K1 enzyme assay, the IC50 of LY-2584702 is 2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.