Learn More
Medchemexpress LLC Ly-266,097 hydrochloride | 172895-39-5 | MFCD00949080 | ≥98.0% | 407.3 g/mol | C21H23ClN2O2 · HCl | 100 MG
SDP
Catalog No. 5000246158
Encompass_Preferred
LY266097 hydrochloride is the hydrochloride salt of LY266097, a selective antagonist of the serotonin 5-HT2B receptor used as a research tool in pharmacology and structural biology studies.
  • Selective 5-HT2B receptor antagonist for receptor pharmacology studies.
  • Supplied as the hydrochloride salt for improved solubility and handling.
  • High purity suitable for in vitro and structural biology applications.
  • Available in small research quantities for assay development and screening.
Catalog No. 50-002-46158 Supplier Medchemexpress LLC Supplier No. HY103094100MG
LY266097 hydrochloride is the hydrochloride salt of LY266097, a selective antagonist of the serotonin 5-HT2B receptor used as a research tool in pharmacology and structural biology studies.
  • Selective 5-HT2B receptor antagonist for receptor pharmacology studies.
  • Supplied as the hydrochloride salt for improved solubility and handling.
  • High purity suitable for in vitro and structural biology applications.
  • Available in small research quantities for assay development and screening.