Learn More
Medchemexpress LLC Ly-272015 hydrochloride | 172895-15-7 | 99.5% | 372.89 | C21H25ClN2O2 | 25 MG
SDP
Catalog No. 5000211282
Encompass_Preferred
LY-272015 hydrochloride is the hydrochloride salt of LY-272015, an orally active and selective 5-HT2B receptor antagonist supplied as a high-purity solid for laboratory research. It is used to probe serotonergic signaling and downstream ERK2 phosphorylation in preclinical studies.
  • Orally active, selective 5-HT2B receptor antagonist.
  • Useful for research into serotonergic signaling and ERK2 phosphorylation.
  • High purity (about 99.5%) for reliable experimental results.
  • Off-white to light yellow solid, suitable for analytical handling.
  • Stable when stored sealed at 4°C; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in small research quantities, including 25 MG.
Catalog No. 50-002-11282 Supplier Medchemexpress LLC Supplier No. HY100851A25MG
LY-272015 hydrochloride is the hydrochloride salt of LY-272015, an orally active and selective 5-HT2B receptor antagonist supplied as a high-purity solid for laboratory research. It is used to probe serotonergic signaling and downstream ERK2 phosphorylation in preclinical studies.
  • Orally active, selective 5-HT2B receptor antagonist.
  • Useful for research into serotonergic signaling and ERK2 phosphorylation.
  • High purity (about 99.5%) for reliable experimental results.
  • Off-white to light yellow solid, suitable for analytical handling.
  • Stable when stored sealed at 4°C; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in small research quantities, including 25 MG.