Learn More
Medchemexpress LLC LY-272015 hydrochloride | 172895-15-7 | 99.5% | 372.89 | C21H25ClN2O2 | 5 MG
SDP
Catalog No. 5000211490
Encompass_Preferred
LY-272015 hydrochloride is a research compound used as a selective 5-HT2B (serotonin) receptor antagonist for preclinical pharmacology studies. It is orally active in animal models and has been reported to show antihypertensive effects in DOCA-salt hypertensive rats. The material is supplied as a solid for laboratory use with supporting analytical documentation.
  • Selective 5-HT2B receptor antagonist for target-specific studies.
  • Orally active in preclinical models, useful for in vivo pharmacology.
  • High purity suitable for research applications.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Provided with data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-11490 Supplier Medchemexpress LLC Supplier No. HY100851A5MG
LY-272015 hydrochloride is a research compound used as a selective 5-HT2B (serotonin) receptor antagonist for preclinical pharmacology studies. It is orally active in animal models and has been reported to show antihypertensive effects in DOCA-salt hypertensive rats. The material is supplied as a solid for laboratory use with supporting analytical documentation.
  • Selective 5-HT2B receptor antagonist for target-specific studies.
  • Orally active in preclinical models, useful for in vivo pharmacology.
  • High purity suitable for research applications.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Provided with data sheet, certificate of analysis, and safety data sheet.