Learn More
Medchemexpress LLC LY-272015 (hydrochloride) | 172895-15-7 | 99.5% | 372.89 g·mol⁻¹ | C21H25ClN2O2 | 50 MG
SDP
Catalog No. 5000211480
Encompass_Preferred
LY-272015 hydrochloride is an orally active, research-grade antagonist of the serotonin 5-HT2B receptor used to probe 5-HT2B-mediated signaling. It inhibits ERK2 phosphorylation induced by 5-HT and has demonstrated antihypertensive effects in animal models. Supplied as a hydrochloride salt with high reported purity suitable for in vitro and in vivo research.
  • Selective 5-HT2B receptor antagonism for target validation and pathway studies.
  • Inhibits ERK2 phosphorylation in cell-based assays.
  • Demonstrated activity in in vivo hypertensive models.
  • High reported purity for reproducible experimental results.
  • Available in small pack sizes convenient for research use.
Catalog No. 50-002-11480 Supplier Medchemexpress LLC Supplier No. HY100851A50MG
LY-272015 hydrochloride is an orally active, research-grade antagonist of the serotonin 5-HT2B receptor used to probe 5-HT2B-mediated signaling. It inhibits ERK2 phosphorylation induced by 5-HT and has demonstrated antihypertensive effects in animal models. Supplied as a hydrochloride salt with high reported purity suitable for in vitro and in vivo research.
  • Selective 5-HT2B receptor antagonism for target validation and pathway studies.
  • Inhibits ERK2 phosphorylation in cell-based assays.
  • Demonstrated activity in in vivo hypertensive models.
  • High reported purity for reproducible experimental results.
  • Available in small pack sizes convenient for research use.