Learn More
Medchemexpress LLC Ly-272015 hydrochloride | 172895-15-7 | MFCD00933882 | 99.5% | 372.89 g/mol | C21H25ClN2O2 | 10 MG
SDP
Catalog No. 5000212356
Encompass_Preferred
LY-272015 hydrochloride is the hydrochloride salt of LY-272015, a selective, orally active 5-HT2B receptor antagonist. It blocks 5-HT or BW723C86-induced ERK2 phosphorylation and has demonstrated antihypertensive activity in DOCA-salt hypertensive rat models. Supplied as a high-purity off-white to light-yellow solid for research use.
  • Selective 5-HT2B receptor antagonist.
  • Orally active compound suitable for in vivo studies.
  • Inhibits 5-HT or BW723C86-induced ERK2 phosphorylation.
  • Demonstrated antihypertensive effects in DOCA-salt hypertensive rats.
  • High reported purity (99.5%).
  • Soluble in DMSO at 5 mg/mL with ultrasonic assistance.
Catalog No. 50-002-12356 Supplier Medchemexpress LLC Supplier No. HY100851A10MG
LY-272015 hydrochloride is the hydrochloride salt of LY-272015, a selective, orally active 5-HT2B receptor antagonist. It blocks 5-HT or BW723C86-induced ERK2 phosphorylation and has demonstrated antihypertensive activity in DOCA-salt hypertensive rat models. Supplied as a high-purity off-white to light-yellow solid for research use.
  • Selective 5-HT2B receptor antagonist.
  • Orally active compound suitable for in vivo studies.
  • Inhibits 5-HT or BW723C86-induced ERK2 phosphorylation.
  • Demonstrated antihypertensive effects in DOCA-salt hypertensive rats.
  • High reported purity (99.5%).
  • Soluble in DMSO at 5 mg/mL with ultrasonic assistance.