Learn More
AdipoGen LY-294,002 (5 mg)
SDP
Supplier:  AdipoGen AGCR10108M005
Encompass_Preferred
Chemical. CAS: 154447-36-6. Formula: C19H17NO3. MW: 307.4. Potent, cell permeable, highly specific, reversible PI(3)K (phosphoinositide 3-kinase) inhibitor. pan-class I / II / III PI3K inhibitor. Acts on the ATP-binding site of the enzyme. In solution more stable than wortmannin. Antagonizes P-glycoprotein-mediated multidrug resistance. Blocks Akt phosphorylation. Pim-1 kinase inhibitor. Autophagy inhibitor (autophagosome formation). Inhibitor of BET bromodomain proteins BRD2, BRD3 and BRD4.
Catalog No. 50-205-3833