Learn More
Apexbio Technology LLC LY 294002, 10mM (in 1mL DMSO). Cas: 154447-36-6 MFCD: MFCD00270881. Potent PI3K inhibitor
SDP
Supplier:  Apexbio Technology LLC A82505.1
Encompass_Preferred
LY294002 is a potent inhibitor of class I phosphoinositide 3-kinases (PI3Ks) [1, 2]. It is cell permeable and reversible, and selective against p110, p110, p110 and p110, which acts on ATP binding site of the catalytic subunit.PI3K family is divided into 3 classes: class I, II and III. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5091