Learn More
Abcam LY 294002 (DMSO solution), PI3-kinase inhibitor, 5MG-LQD
Catalog No. 502704562
Encompass_Preferred
MW 307.3 Da, Purity >99%. A highly selective inhibitor of phosphatidylinositol 3-kinase (IC₅₀ = 1.4 μM). Selective over a range of other kinases including protein kinase C, protein kinase A, MAPK and PI4- kinase (IC₅₀ > 50 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4562 Supplier Abcam Supplier No. AB1465935MGLQD
Add to Cart
Add to Cart
MW 307.3 Da, Purity >99%. A highly selective inhibitor of phosphatidylinositol 3-kinase (IC₅₀ = 1.4 μM). Selective over a range of other kinases including protein kinase C, protein kinase A, MAPK and PI4- kinase (IC₅₀ > 50 μM).
The product is subject to the following: Abcam Restricted Use Statement