Learn More
TARGETMOL CHEMICALS INC LY-294002 HYDROCHLORIDE 25MG
SDP
Catalog No. 502579971
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY-294002 hydrochloride (NSC 697286) is a synthetic molecule inhibitor of PI3K(alpha)/(delta)/(beta) (IC50 0.5/0.57/0.97 uM in cell-free assays); more stable than Wortmannin in solution and also is a blocker of autophagosome formation. purity: 99%
Catalog No. 50-257-9971 Supplier TARGETMOL CHEMICALS INC Supplier No. T407925MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY-294002 hydrochloride (NSC 697286) is a synthetic molecule inhibitor of PI3K(alpha)/(delta)/(beta) (IC50 0.5/0.57/0.97 uM in cell-free assays); more stable than Wortmannin in solution and also is a blocker of autophagosome formation. purity: 99%