Learn More
Medchemexpress LLC HY-114452 10mg Medchemexpress, LY-2940094 CAS:1307245-86-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11445210MG
Encompass_Preferred
Medchemexpress, HY-114452 10mg LY-2940094 CAS:1307245-86-8 LY-2940094 is a potent, selective and orally available nociceptin receptor (NOP receptor) antagonist with high affinity (Ki=0.105 nM) and antagonist potency (Kb=0.166 nM). LY-2940094 reduces ethanol self-administration in animal models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9359