Learn More
Enzo Life Sciences LY-303,511 (1mg). CAS: 154447-38-8
SDP
Supplier:  Enzo Life Sciences ALX270410M001
Encompass_Preferred
Useful negative control for the PI(3)K (phosphoinositide 3-kinase) inhibitor LY-294,002 (BML-ST420). Does not inhibit PI(3)K even at concentrations Alternative name: 2-(4-Piperazinyl)-8-phenyl-4H-1-benzopyran-4-one. Purity: ≥95% (HPLC). Appearance: Yellow solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8537