Learn More
Abcam LY 303511 dihydrochloride, Negative control for LY 294002, 5MG
Catalog No. 502707090
Encompass_Preferred
MW 379.29 g/mol, Purity >98%. Negative control for LY 294002 (ab120243). PI3K inhibitory activity. Inhibits voltage-gated Kv channel activity (IC₅₀ = 64.6 μM). Various pharmacological effects including inhibition of LPS-stimulated FPR1 expression and induction of H₂O₂ production.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7090 Supplier Abcam Supplier No. AB1209565MG
MW 379.29 g/mol, Purity >98%. Negative control for LY 294002 (ab120243). PI3K inhibitory activity. Inhibits voltage-gated Kv channel activity (IC₅₀ = 64.6 μM). Various pharmacological effects including inhibition of LPS-stimulated FPR1 expression and induction of H₂O₂ production.
The product is subject to the following: Abcam Restricted Use Statement