Learn More
Medchemexpress LLC Ly 303511 hydrochloride | 2070014-90-1 | 98.9% | 342.82 g·mol⁻¹ | C19H19ClN2O2 | 10 MG
SDP
Catalog No. 5000287779
Encompass_Preferred
LY 303511 hydrochloride is a research-grade small molecule (CAS 2070014-90-1) that acts as a structural analogue of LY294002; it enhances TRAIL sensitivity in SHEP-1 neuroblastoma cells and reversibly blocks potassium currents in MIN6 insulinoma cells. Supplied as a 10 mg solid with high purity for in vitro studies. Store sealed and protected from moisture.
  • High purity (98.9%) suitable for screening and biochemical assays.
  • Compact 10 mg pack for dose-response and early-stage experiments.
  • Solid form stable under recommended storage temperatures when sealed.
  • Documented activity on TRAIL sensitivity and potassium current modulation.
  • CAS number provided for precise chemical identification.
Catalog No. 50-002-87779 Supplier Medchemexpress LLC Supplier No. HY15643A10MG
LY 303511 hydrochloride is a research-grade small molecule (CAS 2070014-90-1) that acts as a structural analogue of LY294002; it enhances TRAIL sensitivity in SHEP-1 neuroblastoma cells and reversibly blocks potassium currents in MIN6 insulinoma cells. Supplied as a 10 mg solid with high purity for in vitro studies. Store sealed and protected from moisture.
  • High purity (98.9%) suitable for screening and biochemical assays.
  • Compact 10 mg pack for dose-response and early-stage experiments.
  • Solid form stable under recommended storage temperatures when sealed.
  • Documented activity on TRAIL sensitivity and potassium current modulation.
  • CAS number provided for precise chemical identification.