Learn More
Medchemexpress LLC HY-15643A 5mg Medchemexpress, LY 303511 (hydrochloride) CAS:2070014-90-1 Purity:>98%
SDP
Catalog No. 502030659
Encompass_Preferred
Medchemexpress, HY-15643A 5mg LY 303511 (hydrochloride) CAS:2070014-90-1 LY 303511 hydrochloride is a structural analogue of LY294002. LY303511 does not inhibit PI3K. LY303511 enhances TRAIL sensitivity of SHEP-1 neuroblastoma cells. LY303511 reversibly blocks K+ currents (IC50=64.6±9.1 μM) in MIN6 insulinoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0659 Supplier Medchemexpress LLC Supplier No. HY15643A5MG
Medchemexpress, HY-15643A 5mg LY 303511 (hydrochloride) CAS:2070014-90-1 LY 303511 hydrochloride is a structural analogue of LY294002. LY303511 does not inhibit PI3K. LY303511 enhances TRAIL sensitivity of SHEP-1 neuroblastoma cells. LY303511 reversibly blocks K+ currents (IC50=64.6±9.1 μM) in MIN6 insulinoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.