Learn More
Selleck Chemical LLC LY 3200882 5mg 1898283-02-7
SDP
Supplier:  Selleck Chemical LLC S87725MG
Encompass_Preferred
LY 3200882 is a potent, highly selective inhibitor of TGF-? receptor type 1 (TGF?RI). It potently inhibits TGF? mediated SMAD phosphorylation in vitro in tumor and immune cells and in vivo in subcutaneous tumors in a dose dependent fashion. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8312