Learn More
AdipoGen LY-333 531 hydrochloride
SDP
Catalog No. 502562368
Encompass_Preferred
Chemical. CAS 169939-93-9. Formula C28H28N4O3 . HCl. MW 468.6 . 36.5. Isozyme selective inhibitor of protein kinase Cbeta PKCbeta. PKCbetaI and PKCbetaII isozyme inhibitor. Amelioriates diabetic retinopathy, diabetic peripheral neuropathy and diabetic nephropathy. Anti-cancer and anti-angiogenic compound. Suppresses glucose-induced adhesion of human monocytes to endothelial cells. Suppresses ERK1/2 and Akt phosphorylation.
Catalog No. 50-256-2368 Supplier AdipoGen Supplier No. AGCR10087M001
Chemical. CAS 169939-93-9. Formula C28H28N4O3 . HCl. MW 468.6 . 36.5. Isozyme selective inhibitor of protein kinase Cbeta PKCbeta. PKCbetaI and PKCbetaII isozyme inhibitor. Amelioriates diabetic retinopathy, diabetic peripheral neuropathy and diabetic nephropathy. Anti-cancer and anti-angiogenic compound. Suppresses glucose-induced adhesion of human monocytes to endothelial cells. Suppresses ERK1/2 and Akt phosphorylation.