Learn More
Apexbio Technology LLC LY 333531 hydrochloride 169939-93-9 1mg
SDP
Catalog No. 5000062693
Encompass_Preferred
LY 333531 hydrochloride (CAS 169939-93-9) is a biologically active orally available small molecule that selectively inhibits protein kinase C beta (PKC ) It demonstrates ATP-competitive inhibition with reported IC50 values of 4 7 nM for PKC I and 5 9 nM for PKC II indicating high specificity toward these isoforms Due to its targeted inhibition of PKC LY 333531 hydrochloride is extensively studied in the context of diabetic pathology and related complications
Catalog No. 50-000-62693 Supplier Apexbio Technology LLC Supplier No. B77041
LY 333531 hydrochloride (CAS 169939-93-9) is a biologically active orally available small molecule that selectively inhibits protein kinase C beta (PKC ) It demonstrates ATP-competitive inhibition with reported IC50 values of 4 7 nM for PKC I and 5 9 nM for PKC II indicating high specificity toward these isoforms Due to its targeted inhibition of PKC LY 333531 hydrochloride is extensively studied in the context of diabetic pathology and related complications