Learn More
Medchemexpress LLC HY-111540 5mg Medchemexpress, LY-3381916 CAS:2166616-75-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115405MG
Encompass_Preferred
Medchemexpress, HY-111540 5mg LY-3381916 CAS:2166616-75-5 LY-3381916 is a potent, selective and brain penetrated inhibitor of IDO1 activity, binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2371