Learn More
Medchemexpress LLC LY-3381916 (IDO1-IN-5) | 2166616-75-5 | 99.9% | 396.45 | 1 ML
SDP
Catalog No. 5000346972
Encompass_Preferred
LY-3381916 (IDO1-IN-5) is a potent, selective, and brain-penetrated inhibitor of IDO1 activity. It binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1. This product is for research use only and not sold to patients.
  • Potent, selective, and brain-penetrated inhibitor of IDO1 activity.
  • Binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1.
  • No obvious agonism of aryl hydrocarbon receptor (AHR) up to 100 μM.
Catalog No. 50-003-46972 Supplier Medchemexpress LLC Supplier No. HY11154050MG
LY-3381916 (IDO1-IN-5) is a potent, selective, and brain-penetrated inhibitor of IDO1 activity. It binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1. This product is for research use only and not sold to patients.
  • Potent, selective, and brain-penetrated inhibitor of IDO1 activity.
  • Binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1.
  • No obvious agonism of aryl hydrocarbon receptor (AHR) up to 100 μM.