Learn More
American Radiolabeled Chemicals Inc LY 341495-3H 250UC
SDP
Catalog No. 5090105083
Encompass
LY 341495-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 353.37, Shipped in Dry Ice, Exclusive, ((2S)-2-amino-2-((1S,2S)-2-carboxycycloprop-1-yl)-3-xanth-9-yl)propanoic acid)
Catalog No. 50-901-05083 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1439250UC
Add to Cart
Add to Cart
Request a Quote
LY 341495-3H 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 40-60 Ci/mmol, M.W. 353.37, Shipped in Dry Ice, Exclusive, ((2S)-2-amino-2-((1S,2S)-2-carboxycycloprop-1-yl)-3-xanth-9-yl)propanoic acid)