Learn More
Medchemexpress LLC HY-101943 10mg Medchemexpress, LY 345899 CAS:10538-99-5 Purity:>98%
SDP
Catalog No. 501967929
Encompass_Preferred
Medchemexpress, HY-101943 10mg LY 345899 CAS:10538-99-5 LY 345899, a Folate analog, is a methylene tetrahydrofolate dehydrogenase (MTHFD1; DC301) and MTHFD2 ibhibitor with IC50 values of 96 nM and 663 nM, respectively and a Ki of 0.018 μM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7929 Supplier Medchemexpress LLC Supplier No. HY10194310MG
Medchemexpress, HY-101943 10mg LY 345899 CAS:10538-99-5 LY 345899, a Folate analog, is a methylene tetrahydrofolate dehydrogenase (MTHFD1; DC301) and MTHFD2 ibhibitor with IC50 values of 96 nM and 663 nM, respectively and a Ki of 0.018 μM[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.