Learn More
TARGETMOL CHEMICALS INC LY-364947 25MG
SDP
Catalog No. 502581036
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY-364947 (HTS466284) is a potent ATP-competitive inhibitor of TGF(beta)R-I. purity: 99%
Catalog No. 50-258-1036 Supplier TARGETMOL CHEMICALS INC Supplier No. T204825MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LY-364947 (HTS466284) is a potent ATP-competitive inhibitor of TGF(beta)R-I. purity: 99%