Learn More
Apexbio Technology LLC LY 379268,5mg CAS# 191471-52-0
SDP
Supplier:  Apexbio Technology LLC B70615
Encompass_Preferred
LY 379268 is a highly selective group II mGlu receptor agonist with EC50 value of 2.69 and 4.48 nM for hmGlu2 and hmGlu3, respectively [1].mGluRs is a type of glutamate receptor that is being considered as targets for the therapeutic intervention in neurodegenerative disorders. Activation of Group II mGluRs can protect cultured neurones against excitotoxic damage by inhibiting glutamate release [1].In the CA1 hippocampal cells, LY379268 reduced the ischemia-induced hyperactivity and provided protection after 5-min bilateral carotid artery occlusion (BCAO). Furthermore, before 5-min BCAO, 24- or 48-h pretreatment with LY379268 educed the damage to CA1 hippocampal neurons in a gerbil model [2]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0389