Learn More
Medchemexpress LLC HY-50752 10mg , LY-411575 CAS:209984-57-6 Purity:>98%
SDP
Catalog No. 502261079
Encompass_Preferred
HY-50752 10mg LY-411575 CAS: 209984-57-6 LY-411575 is a potent γ-secretase inhibitor with IC50 of 0.078 nM/0.082 nM (membrane/cell-based), and also inhibits Notch S3 cleavage with IC50 of 0.39 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1079 Supplier Medchemexpress LLC Supplier No. HY5075210MG
HY-50752 10mg LY-411575 CAS: 209984-57-6 LY-411575 is a potent γ-secretase inhibitor with IC50 of 0.078 nM/0.082 nM (membrane/cell-based), and also inhibits Notch S3 cleavage with IC50 of 0.39 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.