Learn More
Medchemexpress LLC LY-411575 | 209984-57-6 | 98.7% | C26H23F2N3O4 | 50 MG
SDP
Catalog No. 5000358340
Encompass_Preferred
LY-411575 is a potent γ-secretase inhibitor exhibiting high purity and significant inhibitory activity in both membrane-based and cell-based assays. It also effectively inhibits Notch S3 cleavage, making it suitable for various research applications.
  • Potent gamma-secretase inhibitor with low nanomolar IC50 values.
  • High purity of 98.68%.
  • Effectively inhibits Notch S3 cleavage.
  • Available in solid form for various research needs.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping at room temperature if less than 2 weeks.
Catalog No. 50-003-58340 Supplier Medchemexpress LLC Supplier No. HY5075250MG
LY-411575 is a potent γ-secretase inhibitor exhibiting high purity and significant inhibitory activity in both membrane-based and cell-based assays. It also effectively inhibits Notch S3 cleavage, making it suitable for various research applications.
  • Potent gamma-secretase inhibitor with low nanomolar IC50 values.
  • High purity of 98.68%.
  • Effectively inhibits Notch S3 cleavage.
  • Available in solid form for various research needs.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Shipping at room temperature if less than 2 weeks.