Learn More
Medchemexpress LLC HY-50752 100mg , LY-411575 CAS:209984-57-6 Purity:98%
SDP
Catalog No. 501492945
Encompass_Preferred
Medchemexpress, HY-50752 100mg LY-411575 CAS:209984-57-6 Formula:C26H23F2N3O4 IC50: 0.078 nM (γ-secretase in membrane), 0.082 nM (γ-secretase cell-based), 0.39 nM (Notch S3 cleavage cell-based) Purity:98% LY-411575 is a potent γ-secretase inhibitor with IC 50 of 0.078 nM/0.082 nM (membrane/cell-based), and also inhibits Notch S3 cleavage with IC 50 of 0.39 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2945 Supplier Medchemexpress LLC Supplier No. HY50752100MG
Medchemexpress, HY-50752 100mg LY-411575 CAS:209984-57-6 Formula:C26H23F2N3O4 IC50: 0.078 nM (γ-secretase in membrane), 0.082 nM (γ-secretase cell-based), 0.39 nM (Notch S3 cleavage cell-based) Purity:98% LY-411575 is a potent γ-secretase inhibitor with IC 50 of 0.078 nM/0.082 nM (membrane/cell-based), and also inhibits Notch S3 cleavage with IC 50 of 0.39 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.